Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LPAR3
- Entrez Gene ID
-
23566
- Gene Name
- lysophosphatidic acid receptor 3
- Gene Aliases
-
EDG7, Edg-7, GPCR, HOFNH30, LP-A3, LPA3, RP4-678I3
- Chromosome Location
-
Chr.1: 84813403 - 84893213 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK091731.1 |
- |
1 - 1 |
956 |
82 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CD103667.1 |
- |
1 |
619 |
|
Chr. 1 - 84812061 - 84812765 [-] on Build GRCh38 |
82 |
GenBankmRNA |
BU932493.1 |
- |
|
185 |
|
Chr. - - [-] on Build |
82 |
GenBankmRNA |
BM999782.1 |
- |
1 |
|
|
Chr. 1 - 84811602 - 84812297 [+] on Build GRCh38 |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SSX family member 2 interacting protein |
SSX2IP |
ADIP |
-120.8kb
|
Sense |
Chr. - 84643707 - 84690757 on Build GRCh38 |
Downstream |
mucolipin 2 |
MCOLN2 |
TRP-ML2, TRPML2 |
112.5kb
|
Sense |
Chr. - 84925583 - 84997122 on Build GRCh38 |
View Full Product Details