Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- SUPT3H
- Entrez Gene ID
-
8464
- Gene Name
- SPT3 homolog, SAGA and STAGA complex component
- Gene Aliases
-
SPT3, SPT3L
- Chromosome Location
-
Chr.6: 44808844 - 45378051 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CB133874.1 |
- |
1 |
305 |
|
Chr. 6 - 44827075 - 44827679 [-] on Build GRCh38 |
94 |
GenBankmRNA |
DB326177.1 |
- |
1 |
|
|
Chr. 6 - 44827103 - 44827647 [+] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC105375075 |
LOC105375075 |
|
-275.1kb
|
Sense |
Chr. - 44546161 - 44551881 on Build GRCh38 |
Downstream |
microRNA 586 |
MIR586 |
MIRN586, hsa-mir-586 |
368.4kb
|
Sense |
Chr. - 45197674 - 45197770 on Build GRCh38 |
View Full Product Details