Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CD3D
- Entrez Gene ID
-
915
- Gene Name
- CD3d molecule
- Gene Aliases
-
CD3-DELTA, IMD19, T3D
- Chromosome Location
-
Chr.11: 118339074 - 118344233 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK093775.1 |
- |
1 - 1 |
1248 |
84 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
CD3e molecule |
CD3E |
IMD18, T3E, TCRE |
-26.0kb
|
Anti-sense |
Chr. - 118304580 - 118316175 on Build GRCh38 |
Downstream |
CD3g molecule |
CD3G |
CD3-GAMMA, IMD17, T3G |
47.0
|
Anti-sense |
Chr. - 118344307 - 118353782 on Build GRCh38 |
View Full Product Details