Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK094142.1 - 1 1849 Chr. 2 - 9878460 - 9881335 [+] on Build GRCh38 100


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC100996549 LOC100996549 -99.9kb Anti-sense Chr. - 9758590 - 9778564 on Build GRCh38
Downstream grainyhead like transcription factor 1 GRHL1 LBP32, MGR, NH32, TFCP2L2 70.3kb Sense Chr. - 9951663 - 10002284 on Build GRCh38

View Full Product Details