Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW978145.1 - 1 125 Chr. 19 - 11849061 - 11849323 [+] on Build GRCh38 89
GenBankmRNA DA307070.1 - 1 431 Chr. 19 - 11848750 - 11866220 [+] on Build GRCh38 89
GenBankmRNA DA729391.1 - 1 433 Chr. 19 - 11848748 - 11849495 [+] on Build GRCh38 89
GenBankmRNA BQ424701.1 - 2 79 Chr. 19 - 11848770 - 11867732 [+] on Build GRCh38 89
GenBankmRNA AI870934.1 - 1 102 Chr. 19 - 11849079 - 11851030 [+] on Build GRCh38 89
GenBankmRNA AK094739.1 - 1 455 Chr. 19 - 11848726 - 11851110 [+] on Build GRCh38 89
GenBankmRNA DA934757.1 - 1 433 Chr. 19 - 11848748 - 11849283 [+] on Build GRCh38 89
GenBankmRNA AW206752.1 - 1 119 Chr. 19 - 11849062 - 11867332 [+] on Build GRCh38 89
GenBankmRNA BE219111.1 - 1 120 Chr. 19 - 11849061 - 11866591 [+] on Build GRCh38 89
GenBankmRNA DA167167.1 - 1 435 Chr. 19 - 11848746 - 11849283 [+] on Build GRCh38 89
GenBankmRNA DA948329.1 - 1 435 Chr. 19 - 11848747 - 11849320 [+] on Build GRCh38 89


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream zinc finger protein 440 ZNF440 -13.1kb Sense Chr. - 11814241 - 11835617 on Build GRCh38
Downstream zinc finger protein 69 ZNF69 Cos5, hZNF3 36.7kb Sense Chr. - 11887773 - 11980222 on Build GRCh38

View Full Product Details