Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CA314350.1 - 1 Chr. 17 - 38796177 - 38796856 [+] on Build GRCh38 83
GenBankmRNA AA532745.1 - 1 Chr. 17 - 38796177 - 38796697 [+] on Build GRCh38 83
GenBankmRNA AK095097.1 - 1 1946 Chr. 17 - 38796172 - 38798371 [-] on Build GRCh38 83
GenBankmRNA DB308393.1 - 1 Chr. 17 - 38796172 - 38796715 [+] on Build GRCh38 83
GenBankmRNA AI273265.1 - 1 Chr. 17 - 38796177 - 38796555 [+] on Build GRCh38 83
GenBankmRNA DB378720.1 - 1 Chr. 17 - 38796179 - 38796582 [+] on Build GRCh38 83
GenBankmRNA AW129584.1 - 1 Chr. 17 - 38796177 - 38796641 [+] on Build GRCh38 83
GenBankmRNA CA423058.1 - 1 Chr. 17 - 38796177 - 38796839 [+] on Build GRCh38 83


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream phosphatidylinositol-5-phosphate 4-kinase type 2 beta PIP4K2B PI5P4KB, PIP5K2B, PIP5KIIB, PIP5KIIbeta, PIP5P4KB Sense Chr. - 38765691 - 38799965 on Build GRCh38
Downstream CWC25 spliceosome associated protein homolog CWC25 CCDC49 2.1kb Sense Chr. - 38800434 - 38825350 on Build GRCh38

View Full Product Details