Assay Design
The assay probe spans an exon junction.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR615733.1 |
- |
1 - 2 |
111 |
|
Chr. 5 - 43039399 - 43043170 [-] on Build GRCh38 |
58 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC648987 |
LOC648987 |
|
|
Sense |
Chr. - 43014729 - 43067439 on Build GRCh38 |
Downstream |
uncharacterized LOC100132356 |
LOC100132356 |
|
22.0kb
|
Sense |
Chr. - 43065187 - 43066971 on Build GRCh38 |
View Full Product Details