Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI005389.1 - 1 Chr. 13 - 110894022 - 110894408 [-] on Build GRCh38 66
GenBankmRNA CR617865.1 - 1 1550 Chr. 13 - 110892698 - 110894380 [+] on Build GRCh38 66
GenBankmRNA AV741360.1 - 1 373 Chr. 13 - 110893902 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV724634.1 - 1 395 Chr. 13 - 110893847 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV693186.1 - 1 184 Chr. 13 - 110894065 - 110894418 [+] on Build GRCh38 66
GenBankmRNA H38182.1 - 1 Chr. 13 - 110893995 - 110894405 [-] on Build GRCh38 66
GenBankmRNA BF351536.1 - 1 Chr. 13 - 110893807 - 110894369 [+] on Build GRCh38 67
GenBankmRNA AV724266.1 - 1 475 Chr. 13 - 110893778 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV736826.1 - 1 353 Chr. 13 - 110893902 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AW378651.1 - 1 462 Chr. 13 - 110893786 - 110894373 [+] on Build GRCh38 66
GenBankmRNA AW451227.1 - 1 Chr. 13 - 110894184 - 110894414 [-] on Build GRCh38 66
GenBankmRNA AI653505.1 - 1 Chr. 13 - 110893923 - 110894413 [-] on Build GRCh38 66
GenBankmRNA BX413333.2 - 1 Chr. 13 - 110893911 - 110894380 [-] on Build GRCh38 66
GenBankmRNA BX357497.2 - 1 Chr. 13 - 110893535 - 110894361 [-] on Build GRCh38 66
GenBankmRNA AA682274.1 - 1 Chr. 13 - 110894039 - 110894417 [-] on Build GRCh38 66
GenBankmRNA BI908041.1 - 1 Chr. 13 - 110893674 - 110894338 [-] on Build GRCh38 66
GenBankmRNA AA778203.1 - 1 Chr. 13 - 110894116 - 110894403 [-] on Build GRCh38 66
GenBankmRNA AV738212.1 - 1 275 Chr. 13 - 110893952 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV737027.1 - 1 271 Chr. 13 - 110893980 - 110894418 [+] on Build GRCh38 66
GenBankmRNA DR421641.1 - 1 Chr. 13 - 110893611 - 110894340 [-] on Build GRCh38 66
GenBankmRNA AA825825.1 - 1 Chr. 13 - 110894008 - 110894417 [-] on Build GRCh38 66
GenBankmRNA R98981.1 - Chr. - - [-] on Build 66
GenBankmRNA AV743991.1 - 1 342 Chr. 13 - 110893920 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV738864.1 - 1 173 Chr. 13 - 110894069 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV685355.2 - 1 197 Chr. 13 - 110894047 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AW500094.1 - 1 Chr. 13 - 110894029 - 110894425 [-] on Build GRCh38 66
GenBankmRNA AV737317.1 - 1 287 Chr. 13 - 110893943 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AW074622.1 - 1 Chr. 13 - 110893999 - 110894409 [-] on Build GRCh38 66
GenBankmRNA F02333.1 - Chr. 4 - 91141811 - 91141826 [+] on Build GRCh38 66
GenBankmRNA AI299364.1 - 1 Chr. 13 - 110893971 - 110894403 [-] on Build GRCh38 66
GenBankmRNA AW968041.1 - 1 Chr. 13 - 110893727 - 110894403 [-] on Build GRCh38 66
GenBankmRNA BF003130.1 - 1 Chr. 13 - 110894184 - 110894408 [-] on Build GRCh38 66
GenBankmRNA AV740279.1 - 1 277 Chr. 13 - 110893998 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV686657.1 - 1 191 Chr. 13 - 110894054 - 110894418 [+] on Build GRCh38 66
GenBankmRNA Z39071.1 - Chr. 15 - 79971113 - 79971132 [+] on Build GRCh38 66
GenBankmRNA AV735913.1 - 1 284 Chr. 13 - 110893944 - 110894418 [+] on Build GRCh38 66
GenBankmRNA AV754789.1 - 1 Chr. 13 - 110893855 - 110894392 [-] on Build GRCh38 66
GenBankmRNA CB242408.1 - 1 Chr. 13 - 110893726 - 110894421 [-] on Build GRCh38 66
GenBankmRNA AI653497.1 - 1 Chr. 13 - 110893887 - 110894413 [-] on Build GRCh38 66
GenBankmRNA AV683327.2 - 1 200 Chr. 13 - 110894047 - 110894418 [+] on Build GRCh38 66
GenBankmRNA BE180752.1 - 1 185 Chr. 13 - 110894060 - 110894295 [+] on Build GRCh38 66


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 346 LINC00346 C13orf29, NCRNA00346 -22.4kb Anti-sense Chr. - 110863987 - 110870308 on Build GRCh38
Downstream long intergenic non-protein coding RNA 431 LINC00431 71.6kb Sense Chr. - 110966027 - 110990602 on Build GRCh38

View Full Product Details