Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR624447.1 - 1 2649 Chr. 22 - 46250437 - 46253237 [+] on Build GRCh38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cysteine rich DPF motif domain containing 1 CDPF1 C22orf40 Anti-sense Chr. - 46244013 - 46250679 on Build GRCh38
Downstream polycystin (PKD) family receptor for egg jelly PKDREJ 2.4kb Anti-sense Chr. - 46255663 - 46263322 on Build GRCh38

View Full Product Details