Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CR626528.1 - 1 1143 Chr. 14 - 59629301 - 59630605 [-] on Build GRCh38 96
GenBankmRNA BX408223.2 - 1 399 Chr. 14 - 59629301 - 59629861 [-] on Build GRCh38 96


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream coiled-coil domain containing 175 CCDC175 C14orf38, c14_5395 -47.2kb Sense Chr. - 59504212 - 59577276 on Build GRCh38
Downstream microRNA 5586 MIR5586 16.4kb Sense Chr. - 59646962 - 59647020 on Build GRCh38

View Full Product Details