Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CXXC4
- Entrez Gene ID
-
80319
- Gene Name
- CXXC finger protein 4
- Gene Aliases
-
IDAX
- Chromosome Location
-
Chr.4: 104468306 - 104494967 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
CR738441.1 |
- |
1 - 2 |
177 |
|
Chr. 4 - 104479415 - 104494876 [-] on Build GRCh38 |
162 |
GenBankmRNA |
DA758408.1 |
- |
1 - 2 |
195 |
|
Chr. 4 - 104472087 - 104494894 [-] on Build GRCh38 |
162 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101929468 |
LOC101929468 |
|
|
Anti-sense |
Chr. - 104490965 - 104697592 on Build GRCh38 |
Downstream |
tet methylcytosine dioxygenase 2 |
TET2 |
KIAA1546, MDS |
651.0kb
|
Anti-sense |
Chr. - 105145875 - 105279803 on Build GRCh38 |
View Full Product Details