Product Details

Assay Reference Genome
Location

Chr.13:31211680 on build GRCh38
Cytoband
13q12.3
Species
Homo sapiens
Variation Type
Copy Number

Additional Information

Target Gene Details

Entrez Gene ID
145173
Gene Symbol
B3GLCT
Gene Name
beta 3-glucosyltransferase
Gene Aliases
B3GALTL, B3GTL, B3Glc-T, Gal-T, beta3Glc-T
Location
Chr.13:31199975-31332276 on build GRCh38
Assay Gene Location
Within Intron 3
Gene Symbol Transcript Accession Exon Location Assay Location Protein ID
B3GLCT NM_194318.3 NP_919299.3
XM_006719768.3 XP_006719831.1
XM_011534936.1 XP_011533238.1
XM_017020395.1 XP_016875884.1
AB101481.1 BAD13528.1
AK291273.1
AY190526.1 AAO37647.1
BC068595.1 AAH68595.1
DN999521.1

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2761783 Chr13:31151962 - 31318893 on Build GRCh38 Gain B3GLCT HSPH1
esv2751142 Chr13:31152041 - 31332004 on Build GRCh38 Gain B3GLCT HSPH1
nsv561399 Chr13:31160464 - 31297526 on Build GRCh38 Gain B3GLCT HSPH1
esv3631693 Chr13:31151395 - 31317129 on Build GRCh38 Gain B3GLCT HSPH1
nsv1043630 Chr13:31162792 - 31311663 on Build GRCh38 Gain B3GLCT HSPH1
nsv528568 Chr13:31152516 - 31297526 on Build GRCh38 Gain B3GLCT HSPH1

View Full Product Details