Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KRTAP22-2
Entrez Gene ID
100288287
Gene Name
keratin associated protein 22-2
Gene Aliases
KAP22.2
Chromosome Location
Chr.21: 30590105 - 30590397 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164434.1 NP_001157906.1 1 - 1 143 73
GenBank mRNA AB096950.1 - 1 - 1 143 73

View Full Product Details