Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AA632270.1 - 1 Chr. 7 - 40066389 - 40066881 [-] on Build GRCh38 101
GenBankmRNA AK024379.1 - 1 48 Chr. 7 - 40066704 - 40070406 [+] on Build GRCh38 101
GenBankmRNA BF085735.1 - 1 322 Chr. 7 - 40066430 - 40066819 [+] on Build GRCh38 101
GenBankmRNA AU140620.2 - 1 48 Chr. 7 - 40066704 - 40067486 [+] on Build GRCh38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 265 LINC00265 NCRNA00265 -272.1kb Sense Chr. - 39733568 - 39794623 on Build GRCh38
Downstream M-phase specific PLK1 interacting protein MPLKIP ABHS, C7orf11, ORF20, TTD4 62.3kb Anti-sense Chr. - 40132743 - 40134652 on Build GRCh38

View Full Product Details