Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ELP2
- Entrez Gene ID
-
55250
- Gene Name
- elongator acetyltransferase complex subunit 2
- Gene Aliases
-
SHINC-2, STATIP1, StIP
- Chromosome Location
-
Chr.18: 36129874 - 36177931 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BU517275.1 |
- |
1 |
512 |
|
Chr. 18 - 36176594 - 36177463 [+] on Build GRCh38 |
99 |
GenBankmRNA |
CB149737.1 |
- |
1 |
384 |
|
Chr. 18 - 36176722 - 36177201 [+] on Build GRCh38 |
99 |
GenBankmRNA |
AI547307.1 |
- |
1 |
163 |
|
Chr. 18 - 36176943 - 36177670 [+] on Build GRCh38 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Downstream |
uncharacterized LOC101927809 |
LOC101927809 |
|
2.1kb
|
Anti-sense |
Chr. - 36180001 - 36187435 on Build GRCh38 |
View Full Product Details