Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- TPM4
- Entrez Gene ID
-
7171
- Gene Name
- tropomyosin 4
- Gene Aliases
-
HEL-S-108
- Chromosome Location
-
Chr.19: 16067507 - 16103005 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DW412736.1 |
- |
1 |
151 |
|
Chr. 19 - 16080260 - 16080566 [+] on Build GRCh38 |
103 |
GenBankmRNA |
AW601586.1 |
- |
2 |
|
|
Chr. 19 - 16080056 - 16082037 [-] on Build GRCh38 |
103 |
GenBankmRNA |
DW418909.1 |
- |
1 |
151 |
|
Chr. 19 - 16080260 - 16080655 [+] on Build GRCh38 |
103 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
long intergenic non-protein coding RNA 905 |
LINC00905 |
|
-36.7kb
|
Sense |
Chr. - 16033713 - 16042135 on Build GRCh38 |
Downstream |
RAB8A, member RAS oncogene family |
RAB8A |
MEL, RAB8 |
30.9kb
|
Sense |
Chr. - 16111680 - 16133635 on Build GRCh38 |
View Full Product Details