Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- C7orf34
- Entrez Gene ID
-
135927
- Gene Name
- chromosome 7 open reading frame 34
- Gene Aliases
-
ctm-1
- Chromosome Location
-
Chr.7: 142939506 - 142940870 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AY927625.1 |
- |
1 - 1 |
149 |
104 |
KU178864.1 |
- |
1 - 1 |
392 |
104 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
transient receptor potential cation channel subfamily V member 5 |
TRPV5 |
CAT2, ECAC1, OTRPC3 |
-6.1kb
|
Anti-sense |
Chr. - 142908101 - 142933723 on Build GRCh38 |
Downstream |
Kell blood group, metallo-endopeptidase |
KEL |
CD238, ECE3 |
429.0
|
Anti-sense |
Chr. - 142941114 - 142962416 on Build GRCh38 |
View Full Product Details