The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | BC038560.1 | - | 1 - 2 | 145 | 82 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | microRNA 196a-1 | MIR196A1 | MIRN196-1, MIRN196A1, mir-196a-1 | -72.6kb | Anti-sense | Chr. - 48632490 - 48632559 on Build GRCh38 |
Downstream | prostate cancer susceptibility candidate 2 | PRAC2 | C17orf93, HOXB-AS5, HOXB13-AS1, NCRNA00253 | 13.5kb | Sense | Chr. - 48720802 - 48724758 on Build GRCh38 |