The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001204504.2 | NP_001191433.1 | 30 - 30 | 13564 | 79 |
NM_001204505.2 | NP_001191434.1 | 29 - 29 | 13364 | 79 | |
NM_001204506.2 | NP_001191435.1 | 28 - 28 | 13316 | 79 | |
NM_001297429.1 | NP_001284358.1 | 25 - 25 | 12952 | 79 | |
NM_152910.5 | NP_690874.2 | 29 - 29 | 13527 | 79 | |
NM_178009.4 | NP_821077.1 | 30 - 30 | 13658 | 79 | |
NR_123714.1 | 28 - 28 | 13418 | 79 | ||
GenBank mRNA | AK130015.1 | - | 1 - 1 | 53 | 79 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | VWA8 antisense RNA 1 (head to head) | VWA8-AS1 | -257.5kb | Sense | Chr. - 41961169 - 41981565 on Build GRCh38 | |
Downstream | A-kinase anchoring protein 11 | AKAP11 | AKAP-11, AKAP220, PPP1R44, PRKA11 | 30.5kb | Sense | Chr. - 42271470 - 42323267 on Build GRCh38 |