Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LINC01516
- Entrez Gene ID
-
101929025
- Gene Name
- long intergenic non-protein coding RNA 1516
- Gene Aliases
-
-
- Chromosome Location
-
Chr.10: 25113058 - 25161233 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NR_120649.1 |
|
4 - 5 |
940 |
99 |
GenBank mRNA |
BC042983.2 |
- |
4 - 5 |
940 |
99 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
enkurin, TRPC channel interacting protein |
ENKUR |
C10orf63, CFAP106 |
-62.0kb
|
Anti-sense |
Chr. - 24981979 - 25062279 on Build GRCh38 |
Downstream |
GPR158 antisense RNA 1 |
GPR158-AS1 |
|
32.2kb
|
Anti-sense |
Chr. - 25158072 - 25176276 on Build GRCh38 |
View Full Product Details