Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC043287.1 - 1 2155 Chr. 1 - 94677975 - 94680891 [+] on Build GRCh38 75


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 1057 LINC01057 XLOC_000303 Anti-sense Chr. - 94657533 - 94820281 on Build GRCh38
Downstream microRNA 378g MIR378G 65.0kb Anti-sense Chr. - 94745860 - 94745900 on Build GRCh38

View Full Product Details