Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101928517
Entrez Gene ID
101928517
Gene Name
uncharacterized LOC101928517
Gene Aliases
-
Chromosome Location
Chr.19: 51152923 - 51181966 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_110732.1 2 - 3 541 83
GenBank mRNA BC045766.1 - 2 - 3 541 83


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream sialic acid binding Ig like lectin 7 SIGLEC7 AIRM1, CD328, CDw328, D-siglec, QA79, SIGLEC-7, SIGLEC19P, SIGLECP2, p75, p75/AIRM1 -2.8kb Anti-sense Chr. - 51142261 - 51153526 on Build GRCh38
Downstream sialic acid binding Ig like lectin 17, pseudogene SIGLEC17P HSPC078, SIGLECP2, SIGLECP3 2.1kb Anti-sense Chr. - 51167328 - 51173524 on Build GRCh38

View Full Product Details