Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC048973.1 - 1 945 Chr. 12 - 75623524 - 75625032 [+] on Build GRCh38 72


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream KRR1, small subunit processome component homolog KRR1 HRB2, RIP-1 -111.9kb Anti-sense Chr. - 75497639 - 75511638 on Build GRCh38
Downstream pleckstrin homology like domain family A member 1 PHLDA1 DT1P1B11, PHRIP, TDAG51 400.4kb Anti-sense Chr. - 76025447 - 76031776 on Build GRCh38

View Full Product Details