Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM204A
Entrez Gene ID
63877
Gene Name
family with sequence similarity 204 member A
Gene Aliases
C10orf84, bA319I23.1
Chromosome Location
Chr.10: 118305889 - 118342352 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005270024.1 XP_005270081.1 10 - 10 5983 98
GenBank mRNA BX538256.1 - 1 - 1 2048 98


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BX107735.1 - 1 337 Chr. 10 - 118305887 - 118306356 [-] on Build GRCh38 98
GenBankmRNA AA477431.1 - 1 Chr. 10 - 118305891 - 118306283 [+] on Build GRCh38 98
GenBankmRNA BU785678.1 - 1 Chr. 10 - 118305887 - 118306347 [+] on Build GRCh38 98
GenBankmRNA BU785999.1 - 1 544 Chr. 10 - 118305942 - 118306562 [-] on Build GRCh38 98
GenBankmRNA CK902716.1 - 1 Chr. 10 - 118305887 - 118306505 [+] on Build GRCh38 98
GenBankmRNA AI687652.1 - 1 Chr. 10 - 118305891 - 118306218 [+] on Build GRCh38 98
GenBankmRNA AA479591.1 - 1 336 Chr. 10 - 118305891 - 118306356 [-] on Build GRCh38 98
GenBankmRNA AI740508.1 - 1 Chr. 10 - 118305887 - 118306322 [+] on Build GRCh38 98
GenBankmRNA AI057368.1 - 1 Chr. 10 - 118305887 - 118306115 [+] on Build GRCh38 98
GenBankmRNA AA479290.1 - 1 Chr. 10 - 118305891 - 118306428 [+] on Build GRCh38 98
GenBankmRNA BU630418.1 - 1 Chr. 10 - 118305887 - 118306589 [+] on Build GRCh38 98
GenBankmRNA DB316996.1 - 1 Chr. 10 - 118305891 - 118306420 [+] on Build GRCh38 98
GenBankmRNA BU687362.1 - 1 Chr. 10 - 118305887 - 118306526 [+] on Build GRCh38 98
GenBankmRNA BF109687.1 - 1 Chr. 10 - 118305891 - 118306323 [+] on Build GRCh38 98
GenBankmRNA BU617539.1 - 1 Chr. 10 - 118305887 - 118306323 [+] on Build GRCh38 98
GenBankmRNA DB350494.1 - 1 Chr. 10 - 118305891 - 118306444 [+] on Build GRCh38 98


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cancer susceptibility candidate 2 (non-protein coding) CASC2 C10orf5 -95.7kb Anti-sense Chr. - 118046821 - 118210153 on Build GRCh38
Downstream long intergenic non-protein coding RNA 867 LINC00867 49.0kb Anti-sense Chr. - 118357108 - 118359203 on Build GRCh38

View Full Product Details