Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ADRA1B
- Entrez Gene ID
-
147
- Gene Name
- adrenoceptor alpha 1B
- Gene Aliases
-
ADRA1, ALPHA1BAR
- Chromosome Location
-
Chr.5: 159916733 - 159989205 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BX489252.1 |
- |
1 |
|
|
Chr. 5 - 159955626 - 159956260 [-] on Build GRCh38 |
100 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
uncharacterized LOC101927790 |
LOC101927790 |
|
-12.7kb
|
Anti-sense |
Chr. - 159936619 - 159941617 on Build GRCh38 |
Downstream |
tetratricopeptide repeat domain 1 |
TTC1 |
TPR1 |
52.8kb
|
Sense |
Chr. - 160009100 - 160065545 on Build GRCh38 |
View Full Product Details