Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SESN3
Entrez Gene ID
143686
Gene Name
sestrin 3
Gene Aliases
SEST3
Chromosome Location
Chr.11: 95165513 - 95232541 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271594.1 NP_001258523.1 9 - 9 3818 92
NM_144665.3 NP_653266.2 10 - 10 3916 92
XM_011542614.1 XP_011540916.1 11 - 11 4020 92
GenBank mRNA BX649128.1 - 1 - 1 1156 92


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG259948.1 - 1 560 Chr. 11 - 95170995 - 95171603 [-] on Build GRCh38 92
GenBankmRNA DA217037.1 - 1 86 Chr. 11 - 95170584 - 95171125 [-] on Build GRCh38 92
GenBankmRNA BM454331.1 - 230 Chr. - - [-] on Build 92
GenBankmRNA N78412.1 - 1 88 Chr. 11 - 95170987 - 95171127 [-] on Build GRCh38 92
GenBankmRNA CB321919.1 - 1 104 Chr. 11 - 95170338 - 95171138 [-] on Build GRCh38 92
GenBankmRNA AL707857.1 - 1 378 Chr. 11 - 95170808 - 95171417 [-] on Build GRCh38 92
GenBankmRNA DA189047.1 - 1 414 Chr. 11 - 95170903 - 95171452 [-] on Build GRCh38 92
GenBankmRNA DA757945.1 - 1 446 Chr. 11 - 95170956 - 95171485 [-] on Build GRCh38 92


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream uncharacterized LOC101929295 LOC101929295 -6.4kb Anti-sense Chr. - 95150539 - 95159148 on Build GRCh38
Downstream uncharacterized LOC100129203 LOC100129203 58.0kb Anti-sense Chr. - 95230186 - 95234404 on Build GRCh38

View Full Product Details