Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CPXM2
Entrez Gene ID
119587
Gene Name
carboxypeptidase X (M14 family), member 2
Gene Aliases
CPX2, UNQ676
Chromosome Location
Chr.10: 123745636 - 123944094 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK123463.1 - 1 - 1 647 69


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream G protein-coupled receptor 26 GPR26 -52.4kb Anti-sense Chr. - 123666355 - 123697399 on Build GRCh38
Downstream carbohydrate (N-acetylgalactosamine 4-sulfate 6-O) sulfotransferase 15 CHST15 BRAG, GALNAC4S-6ST 202.9kb Sense Chr. - 124006060 - 124093684 on Build GRCh38

View Full Product Details