Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW838314.1 - 1 81 Chr. 16 - 11008256 - 11008434 [+] on Build GRCh38 121
GenBankmRNA AK124470.1 - 1 2544 Chr. 16 - 11005787 - 11008826 [+] on Build GRCh38 121


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Dexi homolog DEXI MYLE -63.4kb Anti-sense Chr. - 10928891 - 10943007 on Build GRCh38
Downstream suppressor of cytokine signaling 1 SOCS1 CIS1, CISH1, JAB, SOCS-1, SSI-1, SSI1, TIP3 245.6kb Anti-sense Chr. - 11254417 - 11256182 on Build GRCh38

View Full Product Details