Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK124767.1 - 1 3550 Chr. 10 - 25013270 - 25016977 [-] on Build GRCh38 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream enkurin, TRPC channel interacting protein ENKUR C10orf63, CFAP106 Sense Chr. - 24981979 - 25062279 on Build GRCh38
Downstream long intergenic non-protein coding RNA 1516 LINC01516 96.1kb Anti-sense Chr. - 25113058 - 25161233 on Build GRCh38

View Full Product Details