The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AK124984.1 | - | 1 - 1 | 652 | 102 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | SUMO1 activating enzyme subunit 1 | SAE1 | AOS1, HSPC140, SUA1, UBLE1A | -10.2kb | Anti-sense | Chr. - 47130782 - 47210636 on Build GRCh38 |
Downstream | microRNA 3190 | MIR3190 | mir-3190 | 4.4kb | Anti-sense | Chr. - 47226942 - 47227021 on Build GRCh38 |