Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK125205.1 - 1 1950 Chr. 2 - 203326823 - 203329208 [-] on Build GRCh38 122
GenBankmRNA CA418270.1 - 1 60 Chr. 2 - 203292247 - 203327304 [-] on Build GRCh38 122
GenBankmRNA DB317611.1 - 1 Chr. 2 - 203326823 - 203327358 [+] on Build GRCh38 122


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream cytochrome P450 family 20 subfamily A member 1 CYP20A1 CYP-M -21.0kb Anti-sense Chr. - 203238441 - 203305840 on Build GRCh38
Downstream Ras association (RalGDS/AF-6) and pleckstrin homology domains 1 RAPH1 ALS2CR18, ALS2CR9, LPD, PREL-2, PREL2, RMO1, RalGDS/AF-6 104.5kb Sense Chr. - 203433682 - 203535335 on Build GRCh38

View Full Product Details