Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK126341.1 |
- |
1 |
948 |
|
Chr. 13 - 102776596 - 102780565 [-] on Build GRCh38 |
64 |
GenBankmRNA |
DA381525.1 |
- |
1 |
241 |
|
Chr. 13 - 102779244 - 102779859 [-] on Build GRCh38 |
64 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
testis expressed 30 |
TEX30 |
C13orf27 |
-2.8kb
|
Sense |
Chr. - 102765888 - 102773821 on Build GRCh38 |
Downstream |
KDEL motif containing 1 |
KDELC1 |
EP58, ERp58, KDEL1 |
3.7kb
|
Sense |
Chr. - 102784281 - 102799054 on Build GRCh38 |
View Full Product Details