Product Details

Assay Reference Genome
Location

Chr.15:43625350 on build GRCh38
Cytoband
15q15.3
Species
Variation Type
Copy Number

Additional Information

Target Gene Details

Target Copy Number Variation Details

DGV Version
Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv832988 Chr15:43512443 - 43684664 on Build GRCh38 Loss CKMT1B STRC CATSPER2 PPIP5K1 MAP1A
dgv733e212 Chr15:43596009 - 43703698 on Build GRCh38 Gain CKMT1A CKMT1B STRC CATSPER2 STRCP1
nsv437799 Chr15:43491802 - 43697320 on Build GRCh38 Loss TP53BP1 CKMT1A CKMT1B STRC CATSPER2 PPIP5K1 MAP1A
nsv433293 Chr15:43603445 - 43647444 on Build GRCh38 Loss STRC CATSPER2
nsv1514 Chr15:43567062 - 43704382 on Build GRCh38 Deletion CKMT1A CKMT1B STRC CATSPER2 PPIP5K1 STRCP1
dgv2584n100 Chr15:43600874 - 43703678 on Build GRCh38 Gain+Loss CKMT1A STRC CATSPER2 STRCP1
nsv1070270 Chr15:43577801 - 43651202 on Build GRCh38 Deletion CKMT1B STRC CATSPER2 PPIP5K1
dgv2586n100 Chr15:43622539 - 43732292 on Build GRCh38 Loss CKMT1A CATSPER2 STRCP1
esv3892683 Chr15:43623873 - 43698012 on Build GRCh38 Loss CKMT1A CATSPER2
nsv569243 Chr15:43599314 - 43646090 on Build GRCh38 Gain CKMT1B STRC CATSPER2
esv3892682 Chr15:43596778 - 43647444 on Build GRCh38 Loss CKMT1B STRC CATSPER2
nsv817690 Chr15:43622175 - 43647444 on Build GRCh38 Loss CATSPER2
esv33424 Chr15:43596757 - 43703025 on Build GRCh38 Loss CKMT1A CKMT1B STRC CATSPER2 STRCP1
dgv735e212 Chr15:43622429 - 43703698 on Build GRCh38 Loss CKMT1A CATSPER2 STRCP1
nsv569247 Chr15:43601620 - 43647444 on Build GRCh38 Loss STRC CATSPER2
dgv734e212 Chr15:43615418 - 43741783 on Build GRCh38 Loss CATSPER2P1 CKMT1A STRC CATSPER2 STRCP1
nsv1160289 Chr15:43553305 - 43684220 on Build GRCh38 Deletion CKMT1B STRC CATSPER2 PPIP5K1
nsv428304 Chr15:43530960 - 43716382 on Build GRCh38 Loss CKMT1A CKMT1B STRC CATSPER2 PPIP5K1 STRCP1 MAP1A
esv3569487 Chr15:43559201 - 43684983 on Build GRCh38 Gain CKMT1B STRC CATSPER2 PPIP5K1
esv2749626 Chr15:43596837 - 43696748 on Build GRCh38 Deletion CKMT1A CKMT1B STRC CATSPER2
nsv9251 Chr15:43618769 - 43660722 on Build GRCh38 Loss STRC CATSPER2
esv3569520 Chr15:43596064 - 43762536 on Build GRCh38 Gain CATSPER2P1 CKMT1A CKMT1B STRC CATSPER2 STRCP1 PDIA3
dgv732e212 Chr15:43576373 - 43703698 on Build GRCh38 Loss CKMT1A CKMT1B STRC CATSPER2 PPIP5K1 STRCP1
esv24113 Chr15:43559946 - 43696443 on Build GRCh38 Gain+Loss CKMT1A CKMT1B STRC CATSPER2 PPIP5K1
dgv45e203 Chr15:43542120 - 43732292 on Build GRCh38 Gain+Loss CKMT1A CKMT1B STRC CATSPER2 PPIP5K1 STRCP1
esv2421427 Chr15:43592920 - 43696355 on Build GRCh38 Deletion CKMT1A CKMT1B STRC CATSPER2
dgv2583n100 Chr15:43553212 - 43698816 on Build GRCh38 Loss CKMT1A CKMT1B STRC CATSPER2 PPIP5K1
nsv1039949 Chr15:43539681 - 43700448 on Build GRCh38 Gain CKMT1A CKMT1B STRC CATSPER2 PPIP5K1 STRCP1
nsv524910 Chr15:43544280 - 43647444 on Build GRCh38 Loss CKMT1B STRC CATSPER2 PPIP5K1

View Full Product Details