Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BQ185432.1 |
- |
1 |
|
|
Chr. 2 - 24972165 - 24972826 [-] on Build GRCh38 |
97 |
GenBankmRNA |
BQ189107.1 |
- |
|
290 |
|
Chr. - - [-] on Build |
97 |
GenBankmRNA |
AK130860.1 |
- |
1 |
646 |
|
Chr. 2 - 24972104 - 24972821 [+] on Build GRCh38 |
97 |
GenBankmRNA |
BF594769.1 |
- |
1 |
|
|
Chr. 2 - 24972183 - 24972830 [-] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
DNAJC27 antisense RNA 1 |
DNAJC27-AS1 |
|
|
Sense |
Chr. - 24972112 - 25039694 on Build GRCh38 |
Downstream |
EFR3 homolog B |
EFR3B |
KIAA0953 |
69.3kb
|
Sense |
Chr. - 25042104 - 25159137 on Build GRCh38 |
View Full Product Details