Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK025037.1 - 1 2750 Chr. 15 - 77237431 - 77240470 [+] on Build GRCh38 88


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream long intergenic non-protein coding RNA 597 LINC00597 C15orf5 -12.0kb Anti-sense Chr. - 77223893 - 77225404 on Build GRCh38
Downstream high mobility group 20A HMG20A HMGX1, HMGXB1 180.2kb Sense Chr. - 77420651 - 77519899 on Build GRCh38

View Full Product Details