Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA701489.1 |
- |
1 |
|
|
Chr. 7 - 128025627 - 128026016 [-] on Build GRCh38 |
96 |
GenBankmRNA |
W90324.1 |
- |
|
|
|
Chr. - - [-] on Build |
97 |
GenBankmRNA |
AK025082.1 |
- |
1 |
2747 |
|
Chr. 7 - 128023202 - 128026032 [+] on Build GRCh38 |
96 |
GenBankmRNA |
AA886511.1 |
- |
1 |
|
|
Chr. 7 - 128025719 - 128026026 [-] on Build GRCh38 |
96 |
GenBankmRNA |
AA694148.1 |
- |
1 |
|
|
Chr. 7 - 128025605 - 128026016 [-] on Build GRCh38 |
96 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SND1 intronic transcript 1 |
SND1-IT1 |
C7orf54, NAG8, NSG-X |
-23.1kb
|
Sense |
Chr. - 127997509 - 128000077 on Build GRCh38 |
Downstream |
leucine rich repeat containing 4 |
LRRC4 |
NAG14, NGL-2 |
1.0kb
|
Anti-sense |
Chr. - 128027071 - 128032273 on Build GRCh38 |
View Full Product Details