Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK055478.1 - 1 1853 Chr. 18 - 62786567 - 62788941 [+] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream zinc finger CCHC-type containing 2 ZCCHC2 C18orf49 -198.9kb Sense Chr. - 62523410 - 62587709 on Build GRCh38
Downstream BCL2, apoptosis regulator BCL2 Bcl-2, PPP1R50 334.4kb Anti-sense Chr. - 63123346 - 63319778 on Build GRCh38

View Full Product Details