Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- NHSL2
- Entrez Gene ID
-
340527
- Gene Name
- NHS like 2
- Gene Aliases
-
-
- Chromosome Location
-
Chr.X: 71911073 - 72153286 on Build GRCh38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DA682621.1 |
- |
1 |
291 |
|
Chr. X - 72151337 - 72151903 [+] on Build GRCh38 |
97 |
GenBankmRNA |
BE620108.1 |
- |
1 |
205 |
|
Chr. X - 72151422 - 72152229 [+] on Build GRCh38 |
97 |
GenBankmRNA |
CX752026.1 |
- |
1 |
314 |
|
Chr. X - 72151323 - 72152030 [+] on Build GRCh38 |
97 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
TPT1-like protein |
FLJ44635 |
|
|
Sense |
Chr. - 72144184 - 72161750 on Build GRCh38 |
Downstream |
peptidylprolyl cis/trans isomerase, NIMA-interacting 4 |
PIN4 |
EPVH, PAR14, PAR17 |
28.4kb
|
Sense |
Chr. - 72181676 - 72263964 on Build GRCh38 |
View Full Product Details