Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK127858.1 - 1 2747 Chr. 2 - 24284916 - 24288429 [-] on Build GRCh38 102


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream family with sequence similarity 228 member A FAM228A C2orf84 -93.2kb Anti-sense Chr. - 24175043 - 24191698 on Build GRCh38
Downstream nuclear receptor coactivator 1 NCOA1 F-SRC-1, KAT13A, RIP160, SRC1, bHLHe42, bHLHe74 203.6kb Anti-sense Chr. - 24492050 - 24770702 on Build GRCh38

View Full Product Details