Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- UCKL1
- Entrez Gene ID
-
54963
- Gene Name
- uridine-cytidine kinase 1 like 1
- Gene Aliases
-
UCK1L, URKL1
- Chromosome Location
-
Chr.20: 63939829 - 63959435 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK128082.1 |
- |
1 - 1 |
3244 |
71 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB339784.1 |
- |
1 |
|
|
Chr. 20 - 63945307 - 63945861 [-] on Build GRCh38 |
71 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
microRNA 1914 |
MIR1914 |
MIRN1914, hsa-mir-1914 |
-949.0
|
Anti-sense |
Chr. - 63941465 - 63941544 on Build GRCh38 |
Downstream |
UCKL1 antisense RNA 1 |
UCKL1-AS1 |
UCKL1-AS, UCKL1AS, UCKL1OS |
7.5kb
|
Sense |
Chr. - 63953384 - 63956985 on Build GRCh38 |
View Full Product Details