Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AA491220.1 |
- |
1 |
|
|
Chr. 9 - 93075414 - 93075735 [+] on Build GRCh38 |
101 |
GenBankmRNA |
AW971809.1 |
- |
|
|
|
Chr. - - [-] on Build |
101 |
GenBankmRNA |
AK128453.1 |
- |
1 |
2648 |
|
Chr. 9 - 93075435 - 93078209 [-] on Build GRCh38 |
101 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
sushi domain containing 3 |
SUSD3 |
|
|
Anti-sense |
Chr. - 93058688 - 93085138 on Build GRCh38 |
Downstream |
caspase recruitment domain family member 19 |
CARD19 |
BinCARD, C9orf89 |
17.9kb
|
Anti-sense |
Chr. - 93096123 - 93113295 on Build GRCh38 |
View Full Product Details