Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF075065.1 |
- |
2 |
256 |
|
Chr. 8 - 70119878 - 70125628 [-] on Build GRCh38 |
82 |
GenBankmRNA |
R89290.1 |
- |
|
|
|
Chr. - - [-] on Build |
82 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
PR domain 14 |
PRDM14 |
PFM11 |
-48.6kb
|
Sense |
Chr. - 70051126 - 70071327 on Build GRCh38 |
Downstream |
uncharacterized LOC101926892 |
LOC101926892 |
|
350.7kb
|
Anti-sense |
Chr. - 70471134 - 70485687 on Build GRCh38 |
View Full Product Details