Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK023690.1 |
- |
1 |
44 |
|
Chr. 9 - 88177695 - 88179713 [+] on Build GRCh38 |
94 |
GenBankmRNA |
AU139655.1 |
- |
1 |
44 |
|
Chr. 9 - 88177695 - 88178327 [+] on Build GRCh38 |
94 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
SPATA31 subfamily C member 2 |
SPATA31C2 |
FAM75C2 |
-42.7kb
|
Anti-sense |
Chr. - 88129305 - 88134985 on Build GRCh38 |
Downstream |
spindlin 1 |
SPIN1 |
SPIN, TDRD24 |
209.8kb
|
Sense |
Chr. - 88388382 - 88478708 on Build GRCh38 |
View Full Product Details