Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK057714.1 - 1 2251 Chr. 14 - 103057861 - 103061124 [+] on Build GRCh38 101


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream CDC42 binding protein kinase beta CDC42BPB MRCKB -456.0 Anti-sense Chr. - 102932379 - 103057405 on Build GRCh38
Downstream exocyst complex component 3 like 4 EXOC3L4 C14orf73 33.3kb Sense Chr. - 103094202 - 103110562 on Build GRCh38

View Full Product Details