Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK075254.1 - 1 1756 Chr. 9 - 114612512 - 115027100 [+] on Build GRCh38 85


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream transmembrane protein 268 TMEM268 C9orf91 Sense Chr. - 114603897 - 114646423 on Build GRCh38
Downstream uncharacterized LOC100505478 LOC100505478 51.6kb Anti-sense Chr. - 114666434 - 114682089 on Build GRCh38

View Full Product Details