Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
DB314311.1 |
- |
1 |
|
|
Chr. 17 - 7865660 - 7866078 [-] on Build GRCh38 |
69 |
GenBankmRNA |
AK095413.1 |
- |
1 |
2050 |
|
Chr. 17 - 7863790 - 7866078 [+] on Build GRCh38 |
69 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
cytochrome b5 domain containing 1 |
CYB5D1 |
|
-304.0
|
Sense |
Chr. - 7857746 - 7864417 on Build GRCh38 |
Downstream |
chromodomain helicase DNA binding protein 3 |
CHD3 |
Mi-2a, Mi2-ALPHA, ZFH |
18.7kb
|
Sense |
Chr. - 7884764 - 7912757 on Build GRCh38 |
View Full Product Details