Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK124356.1 - 1 1450 Chr. 14 - 93768824 - 93770961 [-] on Build GRCh38 95
GenBankmRNA BU734072.1 - 2 Chr. 14 - 93768806 - 93769581 [+] on Build GRCh38 95


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream proline rich membrane anchor 1 PRIMA1 PRIMA Sense Chr. - 93718298 - 93789029 on Build GRCh38
Downstream FAM181A antisense RNA 1 FAM181A-AS1 C14orf86 133.8kb Sense Chr. - 93904730 - 93926372 on Build GRCh38

View Full Product Details