Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC038707.1 - 1 946 Chr. 22 - 31640507 - 31644129 [-] on Build GRCh38 91


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream microRNA 7109 MIR7109 hsa-mir-7109 -21.4kb Sense Chr. - 31621467 - 31621531 on Build GRCh38
Downstream proline rich 14 like PRR14L C22orf30 37.2kb Sense Chr. - 31681347 - 31750176 on Build GRCh38

View Full Product Details