Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.



Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BC051714.1 - 1 950 Chr. 11 - 47364320 - 47368439 [+] on Build GRCh38 84


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream Spi-1 proto-oncogene SPI1 OF, PU.1, SFPI1, SPI-1, SPI-A Anti-sense Chr. - 47354858 - 47395640 on Build GRCh38
Downstream microRNA 4487 MIR4487 mir-4487 35.3kb Sense Chr. - 47400970 - 47401042 on Build GRCh38

View Full Product Details