Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
A Gene Symbol is not available because the non-coding assay was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID.
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AF119904.1 |
- |
1 |
139 |
|
Chr. 5 - 62261083 - 62534334 [-] on Build GRCh38 |
123 |
GenBankmRNA |
AI207494.1 |
- |
1 |
139 |
|
Chr. 5 - 62533674 - 62534334 [-] on Build GRCh38 |
123 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
importin 11 |
IPO11 |
RanBP11 |
|
Anti-sense |
Chr. - 62412746 - 62628589 on Build GRCh38 |
Downstream |
leucine rich repeat containing 70 |
LRRC70 |
SLRN |
44.4kb
|
Anti-sense |
Chr. - 62578735 - 62581448 on Build GRCh38 |
View Full Product Details